Assay Details
Target Gene Details
Entrez Gene ID: | 226744 |
Gene Name: | consortin, connexin sorting protein |
Gene Aliases: |
9630058J23Rik |
Location: |
Chr.1:179546349-179627473 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnst | NM_146105.3 | 11 | 4126 | NP_666217.2 |
| AK146681.1 | 11 | 4216 | BAE27356.1 | |
| BC034107.1 | 4 | 3144 | AAH34107.1 | |
| BC054802.1 | 11 | 4126 | AAH54802.1 |

Genomic Map