Assay Details
Target Gene Details
Entrez Gene ID: | 226744 |
Gene Name: | consortin, connexin sorting protein |
Gene Aliases: |
9630058J23Rik |
Location: |
Chr.1:179546349-179627473 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnst | NM_146105.3 | 2 | 158 | NP_666217.2 |
| AK036332.1 | 2 | 190 | BAC29388.1 | |
| AK040666.1 | 2 | 181 | BAC30660.1 | |
| AK040899.1 | 2 | 189 | BAC30736.1 | |
| AK087361.1 | 2 | 178 | ||
| AK146681.1 | 2 | 248 | BAE27356.1 | |
| BC054802.1 | 2 | 158 | AAH54802.1 |

Genomic Map