Assay Details
Target Gene Details
Entrez Gene ID: | 226744 |
Gene Name: | consortin, connexin sorting protein |
Gene Aliases: |
9630058J23Rik |
Location: |
Chr.1:179546349-179627473 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnst | NM_146105.3 | 9 | 1774 | NP_666217.2 |
| AK036332.1 | 9 | 1806 | BAC29388.1 | |
| AK146681.1 | 9 | 1864 | BAE27356.1 | |
| BC034107.1 | 2 | 792 | AAH34107.1 | |
| BC054802.1 | 9 | 1774 | AAH54802.1 |

Genomic Map