Assay Details
Target Gene Details
Entrez Gene ID: | 11910 |
Gene Name: | activating transcription factor 3 |
Gene Aliases: |
LRG-21 |
Location: |
Chr.1:191170296-191183333 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atf3 | NM_007498.3 | 4 | 1053 | NP_031524.2 |
| AK133965.1 | 4 | 1057 | BAE21958.1 | |
| AK156786.1 | 4 | 973 | BAE33854.1 | |
| BC019946.1 | 4 | 1064 | AAH19946.1 | |
| BC064799.1 | 4 | 919 | AAH64799.1 | |
| U19118.1 | 4 | 975 | AAB48941.1 |
More Information
Set Membership: |
|

Genomic Map