Assay Details
Target Gene Details
Entrez Gene ID: | 108147 |
Gene Name: | 5-aminoimidazole-4-carboxamide ribonucleotide formyltransferase/IMP cyclohydrolase |
Gene Aliases: |
2610509C24Rik, AA536954, AW212393 |
Location: |
Chr.1:71557156-71579403 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atic | NM_026195.3 | 16 | 2331 | NP_080471.2 |
| AK010611.2 | 16 | 2314 | BAB27060.1 | |
| AK012074.1 | 16 | 2268 | BAB28011.1 | |
| AK076091.1 | 10 | 1623 | BAC36175.1 | |
| BC039925.1 | 16 | 2337 | AAH39925.2 |

Genomic Map