Assay Details
Target Gene Details
Entrez Gene ID: | 68440 |
Gene Name: | dual specificity phosphatase 23 |
Gene Aliases: |
1300005N15Rik, LDP-3 |
Location: |
Chr.1:172630769-172632974 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dusp23 | NM_026725.2 | 1 | 302 | NP_081001.1 |
| AA048085.1 | 1 | 302 | ||
| AB183013.1 | 1 | 290 | BAD26711.1 | |
| AK004912.1 | 1 | 193 | BAB23663.1 | |
| BC069187.1 | 1 | 290 | AAH69187.1 |
More Information
Set Membership: |
|

Genomic Map