Assay Details
Target Gene Details
Entrez Gene ID: | 320404 |
Gene Name: | inositol 1,4,5-trisphosphate 3-kinase B |
Gene Aliases: |
1110033J02Rik, E130307H12Rik |
Location: |
Chr.1:180330476-180424802 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itpkb | NM_001081175.1 | 8 | 4102 | NP_001074644.1 |
| AK039369.1 | 7 | 2017 |
More Information
Set Membership: |
|

Genomic Map