Assay Details
Target Gene Details
Entrez Gene ID: | 11905 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade C (antithrombin), member 1 |
Gene Aliases: |
AI114908, ATIII, At-3, At3 |
Location: |
Chr.1:160978606-161003015 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinc1 | NM_080844.4 | 8 | 1558 | NP_543120.1 |
| AI114908.1 | 1 | 614 | ||
| AK050295.1 | 7 | 1400 | BAC34171.1 | |
| BC019447.1 | 8 | 1518 | AAH19447.1 | |
| BC033377.1 | 7 | 1402 | AAH33377.1 | |
| S47225.1 | 7 | 1361 | AAB23965.1 |
More Information
Set Membership: |
|

Genomic Map