Assay Details
Target Gene Details
Entrez Gene ID: | 81898 |
Gene Name: | splicing factor 3b, subunit 1 |
Gene Aliases: |
155kDa, 2810001M05Rik, AA409119, Prp10, SAP155, SF3b155, TA-8, Targ4 |
Location: |
Chr.1:54985169-55027500 on Build GRCm38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sf3b1 | NM_031179.2 | 23 | 3611 | NP_112456.2 |
| AB037890.1 | 23 | 3636 | BAB40140.1 | |
| AK048521.1 | 1 | 773 | BAC33358.1 | |
| CF952155.1 | 5 | 676 |
More Information
Set Membership: |
|

Genomic Map