Assay Details
Target Gene Details
Entrez Gene ID: | 320460 |
Gene Name: | von Willebrand factor C domain-containing protein 2-like |
Gene Aliases: |
A830006F12Rik, Brl |
Location: |
Chr.1:70725221-70886514 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vwc2l | NM_177164.3 | 2 | 884 | NP_796138.2 |
| AB374230.1 | 1 | 80 | ||
| AK080585.1 | BAC37948.1 | |||
| AK083856.1 | 2 | 848 | BAC39040.1 | |
| BC094541.1 | 2 | 884 | AAH94541.1 | |
| EF552208.1 | 1 | 148 | ||
| EF552209.1 | 1 | 83 | ||
| EF552210.1 |

Genomic Map