Assay Details
Target Gene Details
Entrez Gene ID: | 26361 |
Gene Name: | arginine vasopressin receptor 1B |
Gene Aliases: |
AVPR3, V3/V1b, VIBR, VPR3 |
Location: |
Chr.1:131599114-131612000 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Avpr1b | NM_011924.2 | 2 | 1988 | NP_036054.1 |
| AK033416.1 | 2 | 1022 | ||
| AK050523.1 | 2 | 1070 | BAC34305.1 |
More Information
Set Membership: |
|

Genomic Map