Assay Details
Target Gene Details
Entrez Gene ID: | 26878 |
Gene Name: | UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 2 |
Gene Aliases: |
- |
Location: |
Chr.1:143640697-143649937 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3galt2 | NM_020025.4 | 1 | 107 | NP_064409.3 |
| AK034371.1 | 1 | 63 | BAC28688.1 | |
| AK036141.1 | 1 | 109 | BAC29317.1 | |
| AK080106.1 | ||||
| AK083168.1 | 1 | 62 | BAC38793.1 |
Target Gene Details
Entrez Gene ID: | 214498 |
Gene Name: | cell division cycle 73, Paf1/RNA polymerase II complex component |
Gene Aliases: |
8430414L16Rik, BC027756, C130030P16Rik, C81219, Hrpt2 |
Location: |
Chr.1:143607499-143702888 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdc73 | NM_145991.1 | NP_666103.1 | ||
| AK036714.1 | 1 | 1723 | ||
| AK080861.1 | BAC38048.1 | |||
| BC027756.1 | AAH27756.1 | |||
| BC031127.1 | AAH31127.1 |
More Information
Set Membership: |
|

Genomic Map