Assay Details
Target Gene Details
Entrez Gene ID: | 67446 |
Gene Name: | dual specificity phosphatase 28 |
Gene Aliases: |
0710001B24Rik, AV005521 |
Location: |
Chr.1:92906989-92908620 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dusp28 | NM_175118.3 | 1 | 145 | NP_780327.1 |
| AK002934.1 | 1 | 146 | ||
| AK088940.1 | 1 | 92 | BAC40664.1 | |
| AK163560.1 | 1 | 154 | BAE37397.1 | |
| BC119127.1 | 1 | 134 | ||
| BE652900.1 | 1 | 94 |

Genomic Map