Assay Details
Target Gene Details
Entrez Gene ID: | 67446 |
Gene Name: | dual specificity phosphatase 28 |
Gene Aliases: |
0710001B24Rik, AV005521 |
Location: |
Chr.1:92906989-92908620 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dusp28 | NM_175118.3 | 2 | 784 | NP_780327.1 |
| AK002934.1 | 2 | 783 | ||
| AK088940.1 | 2 | 731 | BAC40664.1 | |
| AK163560.1 | 2 | 793 | BAE37397.1 | |
| BC119127.1 | 2 | 773 |

Genomic Map