Assay Details
Target Gene Details
Entrez Gene ID: | 227289 |
Gene Name: | G protein-coupled bile acid receptor 1 |
Gene Aliases: |
BG37, GPCR, GPR131, M-BAR, TGR5 |
Location: |
Chr.1:74278600-74279589 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpbar1 | NM_174985.1 | 1 | 329 | NP_778150.1 |
| AB086171.1 | 1 | 329 | BAC65344.1 | |
| AB089308.1 | 1 | 329 | BAC55236.1 | |
| BC116914.1 | 1 | 379 | ||
| BC119327.1 | 1 | 379 |
More Information
Set Membership: |
|

Genomic Map