Assay Details
Target Gene Details
Entrez Gene ID: | 98741 |
Gene Name: | potassium voltage gated channel, Shab-related subfamily, member 2 |
Gene Aliases: |
9630047L19Rik, BB130875, Kv2.2 |
Location: |
Chr.1:15287383-15714214 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnb2 | NM_001098528.2 | 2 | 2148 | NP_001091998.1 |
| AK036239.1 | 1 | 39 | ||
| BC146609.1 | 2 | 2148 |
More Information
Set Membership: |
|

Genomic Map