Assay Details
Target Gene Details
Entrez Gene ID: | 16510 |
Gene Name: | potassium voltage-gated channel, subfamily H (eag-related), member 1 |
Gene Aliases: |
EAG1, Kv10.1, M-eag |
Location: |
Chr.1:192190362-192510159 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnh1 | NM_001038607.2 | 9 | 1889 | NP_001033696.1 |
| NM_010600.3 | 9 | 1970 | NP_034730.1 | |
| AK140193.1 | 9 | 1876 | BAE24272.1 | |
| AK147461.1 | 9 | 1883 | BAE27928.1 | |
| BC109012.1 | 9 | 1748 | AAI09013.1 | |
| BC109013.1 | 9 | 1829 | AAI09014.1 | |
| U04294.1 | 9 | 1852 | AAA62474.1 |
More Information
Set Membership: |
|

Genomic Map