Assay Details
Target Gene Details
Entrez Gene ID: | 13603 |
Gene Name: | opsin 3 |
Gene Aliases: |
ERO, Ecpn |
Location: |
Chr.1:175662421-175692902 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Opn3 | NM_010098.3 | 1 | 389 | NP_034228.1 |
| AF140241.1 | 1 | 347 | AAD32670.1 | |
| AK043135.1 | 1 | 390 | BAC31471.1 | |
| AK132822.1 | 1 | 326 | BAE21380.1 | |
| BC104486.1 | 2 | 239 | AAI04487.1 | |
| BC105646.1 | 2 | 186 | ||
| BC105919.1 | 2 | 186 | AAI05920.1 | |
| BC139805.1 | 2 | 187 |
More Information
Set Membership: |
|

Genomic Map