Assay Details
Target Gene Details
Entrez Gene ID: | 71740 |
Gene Name: | poliovirus receptor-related 4 |
Gene Aliases: |
1200017F15Rik, Prr4 |
Location: |
Chr.1:171370173-171388287 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pvrl4 | NM_001122680.1 | 3 | 1080 | NP_001116152.1 |
| NM_027893.3 | 3 | 1080 | NP_082169.2 | |
| AF472510.1 | 3 | 585 | AAL79833.1 | |
| AK004821.1 | 3 | 899 | BAB23592.1 | |
| AK028479.1 | 3 | 856 | BAC25972.1 | |
| AK154983.1 | 4 | 778 | BAE32972.1 | |
| BC024948.1 | 3 | 836 | AAH24948.1 |

Genomic Map