Assay Details
Target Gene Details
Entrez Gene ID: | 71740 |
Gene Name: | poliovirus receptor-related 4 |
Gene Aliases: |
1200017F15Rik, Prr4 |
Location: |
Chr.1:171370173-171388287 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pvrl4 | NM_001122680.1 | 8 | 2572 | NP_001116152.1 |
| NM_027893.3 | 9 | 2647 | NP_082169.2 | |
| AK004821.1 | 8 | 2391 | BAB23592.1 | |
| AK028479.1 | 9 | 2423 | BAC25972.1 | |
| AK154983.1 | 10 | 2345 | BAE32972.1 | |
| BC024948.1 | 9 | 2403 | AAH24948.1 |

Genomic Map