Assay Details
Target Gene Details
Entrez Gene ID: | 14859 |
Gene Name: | glutathione S-transferase, alpha 3 |
Gene Aliases: |
Gst2-3 |
Location: |
Chr.1:21240585-21265575 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gsta3 | NM_001077353.2 | 8 | 1083 | NP_001070821.1 |
| NM_001288617.1 | 8 | 1128 | NP_001275546.1 | |
| NM_010356.4 | 7 | 988 | NP_034486.2 | |
| AI303497.1 | 1 | 399 | ||
| AK002478.1 | BAB22131.1 | |||
| AK014076.1 | BAB29143.1 | |||
| AK149543.1 | 7 | 953 | BAE28948.1 | |
| M73483.1 | AAA37751.1 | |||
| X65021.1 | CAA46155.1 |
More Information
Set Membership: |
|

Genomic Map