Assay Details
Target Gene Details
Entrez Gene ID: | 110611 |
Gene Name: | high density lipoprotein (HDL) binding protein |
Gene Aliases: |
1110005P14Rik, AA960365, AI118566, D1Ertd101e |
Location: |
Chr.1:93405940-93478917 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hdlbp | NM_001301364.1 | 28 | 5105 | NP_001288293.1 |
| NM_133808.5 | 28 | 5188 | NP_598569.1 | |
| BC027788.1 | 28 | 5083 | AAH27788.1 |
More Information
Set Membership: |
|

Genomic Map