Assay Details
Target Gene Details
Entrez Gene ID: | 16526 |
Gene Name: | potassium channel, subfamily K, member 2 |
Gene Aliases: |
A430027H14Rik, AI848635, TREK-1 |
Location: |
Chr.1:189207930-189402705 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnk2 | NM_001159850.1 | 2 | 520 | NP_001153322.1 |
| NM_001281847.1 | 2 | 411 | NP_001268776.1 | |
| NM_001281848.1 | 2 | 370 | NP_001268777.1 | |
| NM_010607.3 | 2 | 616 | NP_034737.2 | |
| AK138427.1 | 2 | 292 | BAE23658.1 | |
| AK140716.1 | 2 | 291 | BAE24455.1 | |
| AK164197.1 | 2 | 370 | ||
| AY736359.1 | 2 | 255 | AAV48996.1 | |
| BC062094.1 | 2 | 234 | AAH62094.1 | |
| BY267214.1 | 1 | 55 | ||
| U73488.2 | 3 | 693 | AAC53005.2 |
More Information
Set Membership: |
|

Genomic Map