Assay Details
Target Gene Details
Entrez Gene ID: | 17681 |
Gene Name: | musculin |
Gene Aliases: |
MyoR, bHLHa22 |
Location: |
Chr.1:14753344-14756012 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msc | NM_010827.2 | 2 | 1556 | NP_034957.1 |
| AF087035.1 | 2 | 1556 | AAC69869.1 | |
| BC103592.1 | AAI03593.1 | |||
| BC103593.1 | AAI03594.1 | |||
| BC103594.1 | AAI03595.1 | |||
| BC103623.1 | AAI03624.1 |
More Information
Set Membership: |
|

Genomic Map