Assay Details
Target Gene Details
Entrez Gene ID: | 72085 |
Gene Name: | O-sialoglycoprotein endopeptidase-like 1 |
Gene Aliases: |
2610001M19Rik, AA416452 |
Location: |
Chr.1:53312897-53326343 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Osgepl1 | NM_001285839.1 | 3 | 737 | NP_001272768.1 |
| NR_104360.1 | 3 | 732 | ||
| NR_104361.1 | 3 | 737 | ||
| AK011265.1 | 3 | 685 | BAB27506.1 | |
| AK045669.1 | 3 | 699 | BAC32450.1 | |
| AK137332.1 | 3 | 688 | BAE23308.1 | |
| AK137402.1 | 2 | 3398 | ||
| BC058172.1 | 3 | 700 | AAH58172.1 |
More Information
Set Membership: |
|

Genomic Map