Assay Details
Target Gene Details
Entrez Gene ID: | 72085 |
Gene Name: | O-sialoglycoprotein endopeptidase-like 1 |
Gene Aliases: |
2610001M19Rik, AA416452 |
Location: |
Chr.1:53312897-53326343 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Osgepl1 | NM_001285839.1 | 4 | 1043 | NP_001272768.1 |
| NR_104360.1 | 4 | 1038 | ||
| NR_104361.1 | 4 | 1043 | ||
| AK011265.1 | 4 | 991 | BAB27506.1 | |
| AK045669.1 | 4 | 1005 | BAC32450.1 | |
| AK137332.1 | 4 | 994 | BAE23308.1 | |
| BC058172.1 | 4 | 1006 | AAH58172.1 |
More Information
Set Membership: |
|

Genomic Map