Assay Details
Target Gene Details
Entrez Gene ID: | 19335 |
Gene Name: | RAB23, member RAS oncogene family |
Gene Aliases: |
AW545388, opb, opb2 |
Location: |
Chr.1:33719896-33742564 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab23 | NM_008999.4 | 1 | 48 | NP_033025.2 |
| AK047068.1 | 1 | 49 | BAC32949.1 | |
| AK054079.1 | 1 | 34 | ||
| AK082017.1 | ||||
| AK163909.1 | 1 | 57 | ||
| AK164057.1 | 1 | 35 | ||
| BC025578.1 | AAH25578.1 |

Genomic Map