Assay Details
Target Gene Details
Entrez Gene ID: | 208618 |
Gene Name: | enhancer trap locus 4 |
Gene Aliases: |
6620402G01Rik, 9430077C05Rik, BC026657, E330027G05Rik, Etl-4, Skt |
Location: |
Chr.2:19909763-20810535 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Etl4 | NM_001081006.1 | 7 | 1224 | NP_001074475.1 |
| NM_001177630.2 | 6 | 1322 | NP_001171101.2 | |
| NM_001177631.1 | 6 | 1322 | NP_001171102.1 | |
| NM_029895.4 | 7 | 1535 | NP_084171.2 | |
| NM_178059.5 | 7 | 1535 | NP_835160.2 | |
| AB093289.2 | 6 | 1112 | BAC41473.2 | |
| AB125594.2 | 7 | 1535 | BAD14929.1 | |
| AB125595.2 | 7 | 1535 | BAD14930.1 | |
| AK052020.1 | 8 | 1637 | BAC34833.1 | |
| AK054453.1 | 2 | 478 | BAC35783.1 | |
| AK086078.1 | 2 | 478 | ||
| AK147510.1 | 6 | 1229 | ||
| AK158471.1 | 2 | 270 | BAE34528.1 | |
| BC158051.1 | 6 | 1044 |

Genomic Map