Assay Details
Target Gene Details
Entrez Gene ID: | 74030 |
Gene Name: | Ras and Rab interactor 2 |
Gene Aliases: |
2010003K16Rik, 4632403N06Rik, AW821980, RASSF4 |
Location: |
Chr.2:145771727-145887616 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rin2 | NM_028724.4 | 13 | 3319 | NP_083000.5 |
| AK014548.1 | 5 | 1791 | BAB29425.1 | |
| AK040763.1 | 5 | 1791 | BAC30697.1 | |
| AK150680.1 | 11 | 3127 | BAE29760.1 | |
| AK151332.1 | 11 | 3083 | BAE30311.1 | |
| AK152025.1 | 6 | 1764 | BAE30885.1 | |
| AK152893.1 | 11 | 3127 | BAE31576.1 | |
| AK154421.1 | 5 | 1735 | BAE32573.1 | |
| AK163047.1 | 5 | 2306 | BAE37170.1 | |
| AK171064.1 | 5 | 1706 | BAE42223.1 | |
| BC005529.1 | 5 | 1752 | AAH05529.1 | |
| BC040390.1 | 13 | 3313 |
More Information
Set Membership: |
|

Genomic Map