Assay Details
Target Gene Details
Entrez Gene ID: | 51902 |
Gene Name: | ring finger protein 24 |
Gene Aliases: |
2810473M14Rik, 4930505A13Rik, AI317164, C86507, D2Ertd504e |
Location: |
Chr.2:131298488-131352892 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf24 | NM_178607.4 | 6 | 1324 | NP_848722.1 |
| AK015699.1 | 7 | 1384 | ||
| AK048813.1 | 6 | 1282 | BAC33467.1 | |
| AK083665.1 | 6 | 1314 | BAC38987.1 | |
| AK089207.1 | 6 | 1311 | BAC40793.1 | |
| AK154093.1 | 6 | 1261 | BAE32370.1 | |
| AK165427.1 | 6 | 1282 | BAE38182.1 | |
| BC096529.1 | 6 | 1257 | AAH96529.1 |

Genomic Map