Assay Details
Target Gene Details
Entrez Gene ID: | 66971 |
Gene Name: | CDK5 regulatory subunit associated protein 1 |
Gene Aliases: |
2310066P17Rik |
Location: |
Chr.2:154289809-154372850 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk5rap1 | NM_025876.2 | 2 | 150 | NP_080152.1 |
| AK010081.1 | 2 | 150 | ||
| AK010097.1 | 2 | 150 | BAB26700.1 | |
| AK088517.1 | 2 | 159 | BAC40398.1 | |
| AK136570.1 | 2 | 163 | BAE23052.1 | |
| BC025132.1 | 1 | 115 | AAH25132.1 |

Genomic Map