Assay Details
Target Gene Details
Entrez Gene ID: | 110911 |
Gene Name: | CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2 |
Gene Aliases: |
5730450N06Rik, 5730460C18Rik, AI854580, D2Wsu127e |
Location: |
Chr.2:132262609-132312050 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cds2 | NM_001291039.1 | 13 | 3474 | NP_001277968.1 |
| NM_138651.7 | 13 | 3797 | NP_619592.1 | |
| AK036328.1 | 13 | 3648 | BAC29384.1 | |
| AK147541.1 | 13 | 3689 | BAE27984.1 | |
| AK166001.1 | 13 | 3648 | BAE38511.1 |
More Information
Set Membership: |
|

Genomic Map