Assay Details
Target Gene Details
Entrez Gene ID: | 98952 |
Gene Name: | family with sequence similarity 102, member A |
Gene Aliases: |
AI426465, C230093N12Rik, Eeig1 |
Location: |
Chr.2:32535359-32569750 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam102a | NM_153560.4 | 1 | 435 | NP_705788.1 |
| AK049032.1 | 1 | 437 | BAC33519.1 | |
| AK131145.1 | 1 | 462 | BAD21395.1 | |
| AK157331.1 | 1 | 65 | BAE34050.1 | |
| AK157490.1 | 1 | 62 | BAE34101.1 | |
| AK158435.1 | 1 | 62 | ||
| AK165751.1 | 1 | 62 | ||
| AK170536.1 | 1 | 463 | BAE41864.1 | |
| AK170847.1 | 1 | 56 | BAE42070.1 | |
| AK170951.1 | 1 | 62 | BAE42135.1 | |
| AK172536.1 | 1 | 62 | BAE43056.1 |

Genomic Map