Assay Details
Target Gene Details
Entrez Gene ID: | 327655 |
Gene Name: | diphosphoinositol pentakisphosphate kinase 1 |
Gene Aliases: |
B430315C20Rik, Hisppd2a, mKIAA0377 |
Location: |
Chr.2:121310557-121355636 on Build GRCm38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppip5k1 | NM_178795.4 | 30 | 4571 | NP_848910.3 |
| AF502585.1 | 30 | 4573 | AAP46293.1 | |
| AK122264.1 | 30 | 4537 | BAC65546.1 | |
| BC065138.1 | 17 | 2803 | AAH65138.1 | |
| BC138351.1 | 30 | 4619 |
More Information
Set Membership: |
|

Genomic Map