Assay Details
Target Gene Details
Entrez Gene ID: | 229279 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A3 |
Gene Aliases: |
2410013L13Rik, 2610209F03Rik, 2610510D13Rik, Hnrpa3 |
Location: |
Chr.2:75659230-75669407 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpa3 | NM_053263.1 | 11 | 2488 | NP_444493.1 |
| NM_146130.3 | 11 | 2554 | NP_666242.2 | |
| NM_198090.2 | NP_932758.1 | |||
| AK011930.1 | 1 | 864 | ||
| AK076236.1 | 1 | 404 | ||
| BC038364.1 | AAH38364.1 | |||
| BC064824.1 | AAH64824.1 | |||
| BC116250.1 | ||||
| BC116251.1 |
More Information
Set Membership: |
|

Genomic Map