Assay Details
Target Gene Details
Entrez Gene ID: | 227618 |
Gene Name: | leucine rich repeat containing 26 |
Gene Aliases: |
- |
Location: |
Chr.2:25289911-25291193 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc26 | NM_146117.2 | 2 | 1046 | NP_666229.1 |
| AK137758.1 | 2 | 1042 | BAE23492.1 | |
| AK142669.1 | 2 | 1034 | BAE25154.1 | |
| BC004853.1 | 2 | 1041 | AAH04853.1 | |
| BC024531.1 | 2 | 1016 | AAH24531.1 | |
| BC024539.1 | 2 | 1010 | AAH24539.1 |

Genomic Map