Assay Details
Target Gene Details
Entrez Gene ID: | 69702 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 1 |
Gene Aliases: |
2410001M24Rik, CGI-65, CIA30 |
Location: |
Chr.2:119655446-119662885 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufaf1 | NM_027175.3 | 5 | 1288 | NP_081451.3 |
| AK010328.1 | 4 | 1118 | BAB26855.2 | |
| AK050343.1 | 5 | 1289 | BAC34201.1 | |
| BC018422.1 | 5 | 1118 | AAH18422.1 |

Genomic Map