Assay Details
Target Gene Details
Entrez Gene ID: | 20014 |
Gene Name: | ribophorin II |
Gene Aliases: |
1300012C06Rik, AV261018, Rpn-2 |
Location: |
Chr.2:157279011-157326318 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpn2 | NM_019642.4 | 17 | 2513 | NP_062616.2 |
| AK004968.1 | 17 | 2514 | BAB23707.1 | |
| AK018761.2 | 17 | 2553 | ||
| AK087923.1 | 11 | 1542 | ||
| AK167864.1 | 17 | 2293 | ||
| AK167866.1 | 17 | 2293 | ||
| AK168221.1 | 17 | 2293 |
More Information
Set Membership: |
|

Genomic Map