Assay Details
Target Gene Details
Entrez Gene ID: | 227522 |
Gene Name: | ribonuclease P/MRP 38 subunit |
Gene Aliases: |
C330006A15Rik |
Location: |
Chr.2:3328943-3333024 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpp38 | NM_001013376.2 | 2 | 412 | NP_001013394.2 |
| AK049133.1 | 2 | 357 | BAC33561.1 | |
| AK087782.1 | 2 | 413 | BAC40000.1 | |
| AK139023.1 | 2 | 340 | BAE23865.1 | |
| BC051448.1 | 2 | 387 | AAH51448.1 |

Genomic Map