Assay Details
Target Gene Details
Entrez Gene ID: | 26569 |
Gene Name: | solute carrier family 27 (fatty acid transporter), member 4 |
Gene Aliases: |
BB144259, FATP4 |
Location: |
Chr.2:29802680-29817522 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc27a4 | NM_011989.4 | 1 | 95 | NP_036119.1 |
| AK036919.1 | 1 | 97 | BAC29639.1 | |
| AK155313.1 | 1 | 65 | ||
| AK155388.1 | 1 | 65 | BAE33236.1 | |
| AK170377.1 | 1 | 99 | BAE41756.1 | |
| AK196043.1 | ||||
| AK217898.1 | ||||
| BC023114.1 | AAH23114.1 |
More Information
Set Membership: |
|

Genomic Map