Assay Details
Target Gene Details
Entrez Gene ID: | 381246 |
Gene Name: | X Kell blood group precursor related family member 9 homolog |
Gene Aliases: |
Gm1620, XRG9 |
Location: |
Chr.1:13668738-13701723 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xkr9 | NM_001011873.2 | 4 | 1119 | NP_001011873.1 |
| AY534255.1 | 4 | 1119 | AAT07104.1 | |
| BC147699.1 | 3 | 725 | ||
| BC158121.1 | 3 | 980 |

Genomic Map