Assay Details
Target Gene Details
Entrez Gene ID: | 227737 |
Gene Name: | family with sequence similarity 129, member B |
Gene Aliases: |
9130404D14Rik |
Location: |
Chr.2:32876134-32925253 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam129b | NM_146119.2 | 14 | 3381 | NP_666231.1 |
| AK033735.1 | 14 | 3349 | BAC28456.1 | |
| AK046583.1 | 14 | 3278 | BAC32795.1 | |
| AK051396.1 | 14 | 3320 | BAC34625.1 | |
| AK146953.1 | 14 | 3366 | BAE27562.1 | |
| AK149818.1 | ||||
| AK149888.1 | 14 | 3291 | BAE29147.1 | |
| AK150642.1 | 14 | 3319 | BAE29730.1 | |
| AK152207.1 | 14 | 3340 | BAE31035.1 | |
| AK153489.1 | 14 | 3325 | BAE32037.1 | |
| AK170512.1 | 14 | 3321 | BAE41849.1 | |
| AK195145.1 | 1 | 199 | ||
| BC024639.1 | 6 | 2114 | AAH24639.1 | |
| BC027843.1 | 14 | 3301 | AAH27843.1 |

Genomic Map