Assay Details
Target Gene Details
Entrez Gene ID: | 68222 |
Gene Name: | family with sequence similarity 166, member A |
Gene Aliases: |
4931415M17 |
Location: |
Chr.2:25214724-25222281 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam166a | NM_026624.3 | 3 | 402 | NP_080900.1 |
| AK016459.1 | 3 | 404 | BAB30248.1 | |
| BC061039.1 | 3 | 405 | AAH61039.1 |

Genomic Map