Assay Details
Target Gene Details
Entrez Gene ID: | 15434 |
Gene Name: | homeobox D3 |
Gene Aliases: |
Hox-4.1, Hox-5.5 |
Location: |
Chr.2:74711993-74748271 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hoxd3 | NM_010468.2 | 1 | 218 | NP_034598.2 |
| AK138525.1 | 1 | 219 | BAE23694.1 |
Target Gene Details
Entrez Gene ID: | 73429 |
Gene Name: | homeobox D3, opposite strand 1 |
Gene Aliases: |
1700064D17Rik, 1700109F18Rik |
Location: |
Chr.2:74710044-74716130 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hoxd3os1 | NR_027899.1 | |||
| AK007148.1 | ||||
| AK054396.1 | 1 | 317 | BAC35762.1 | |
| AK164120.1 | 1 | 219 |
More Information
Set Membership: |
|

Genomic Map