Assay Details
Target Gene Details
Entrez Gene ID: | 15434 |
Gene Name: | homeobox D3 |
Gene Aliases: |
Hox-4.1, Hox-5.5 |
Location: |
Chr.2:74711993-74748271 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hoxd3 | NM_010468.2 | 2 | 842 | NP_034598.2 |
| AK133705.1 | 2 | 710 | BAE21791.1 | |
| AK138525.1 | 2 | 843 | BAE23694.1 | |
| BC137716.1 | 1 | 467 | ||
| BC137718.1 | 1 | 467 | ||
| U56400.1 | 1 | 518 | AAC52779.1 | |
| X73573.1 | 1 | 507 | CAA51975.1 |

Genomic Map