Assay Details
Target Gene Details
Entrez Gene ID: | 15434 |
Gene Name: | homeobox D3 |
Gene Aliases: |
Hox-4.1, Hox-5.5 |
Location: |
Chr.2:74711993-74748271 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hoxd3 | NM_010468.2 | NP_034598.2 | ||
| AK138525.1 | BAE23694.1 |
Target Gene Details
Entrez Gene ID: | 15436 |
Gene Name: | homeobox D4 |
Gene Aliases: |
6030436D05Rik, Hox-4.2, Hox-5.1 |
Location: |
Chr.2:74721978-74729160 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hoxd4 | NM_010469.2 | 5 | 1106 | NP_034599.2 |
| BC139206.1 | ||||
| BC139207.1 | ||||
| J03770.1 | 1 | 77 | AAA20072.1 |
More Information
Set Membership: |
|

Genomic Map