Assay Details
Target Gene Details
Entrez Gene ID: | 241528 |
Gene Name: | leucine rich repeat containing 55 |
Gene Aliases: |
A330097E02Rik, Gm351 |
Location: |
Chr.2:85188067-85198932 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc55 | NM_001033346.2 | 1 | 354 | NP_001028518.1 |

Genomic Map