Assay Details
Target Gene Details
Entrez Gene ID: | 111368 |
Gene Name: | prion protein gene complex |
Gene Aliases: |
Prnp-Prnd |
Location: |
Chr.2:131909928-131956131 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prn | NM_001278258.1 | 4 | 795 | NP_001265187.1 |
| NM_001278259.1 | 4 | 795 | NP_001265188.1 |
Target Gene Details
Entrez Gene ID: | 26434 |
Gene Name: | prion protein dublet |
Gene Aliases: |
AI450264, Dpl, PrPLP, doppel |
Location: |
Chr.2:131950861-131956131 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prnd | NM_001126338.2 | 2 | 573 | NP_001119810.1 |
| NM_001278257.1 | 2 | 573 | NP_001265186.1 | |
| NM_001278520.1 | 2 | 552 | NP_001265449.1 | |
| NM_023043.3 | 2 | 573 | NP_075530.1 | |
| AF165165.1 | 2 | 552 | AAF02544.1 | |
| AF165166.1 | 2 | 556 | AAF02545.1 | |
| AF192384.1 | 2 | 572 | AAF09198.1 | |
| AF192385.1 | 2 | 572 | AAF09199.1 | |
| AK043023.1 | 2 | 574 | BAC31436.1 | |
| AK161043.1 | 2 | 574 | BAE36166.1 | |
| BC025140.1 | 2 | 549 | AAH25140.1 | |
| DQ832281.1 | 2 | 574 | ||
| GQ245668.1 |

Genomic Map