Assay Details
Target Gene Details
Entrez Gene ID: | 68295 |
Gene Name: | AAR2 splicing factor homolog (S. cerevisiae) |
Gene Aliases: |
0610011L14Rik |
Location: |
Chr.2:156547558-156568972 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aar2 | NM_001164818.1 | 1 | 255 | NP_001158290.1 |
| NM_026661.4 | 1 | 255 | NP_080937.3 | |
| AK018741.1 | 1 | 236 | BAB31382.1 | |
| AK028890.1 | 1 | 233 | BAC26177.1 | |
| AK166203.1 | 1 | 236 | BAE38626.1 | |
| AK169526.1 | 1 | 247 | BAE41210.1 | |
| AK187905.1 | 1 | 119 | ||
| AK200377.1 | 1 | 117 | ||
| AK201919.1 | 1 | 118 | ||
| AK210506.1 | 1 | 117 | ||
| BC019988.1 | 1 | 179 | AAH19988.1 | |
| BC020112.1 | 1 | 153 | AAH20112.1 | |
| CJ152371.1 | 1 | 256 |
More Information
Set Membership: |
|

Genomic Map