Assay Details
Target Gene Details
Entrez Gene ID: | 22625 |
Gene Name: | mitogen-activated protein kinase kinase kinase 19 |
Gene Aliases: |
Ysk4 |
Location: |
Chr.1:127814962-127855613 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map3k19 | NM_011737.1 | 9 | 3635 | NP_035867.1 |

Genomic Map